Filter centering rings for small flange connections
Supplier: VACUUBRAND
20660160 20660161 20660162 20660163
185-1291EA 0 INR
185-1291 185-1292 185-1293 185-1294
Filter centering rings for small flange connections
Seals Sealing Rings
These filter centering rings for small flange KF connections made of stainless steel with NBR sealing ring.