Diaphragm pumps
Supplier: VACUUBRAND
20696081 20696082 696434 20738000 20731102 20730000 20730002 20743002 20743001 20741002 20741000 20743000 20744000
181-1557EA 0 INR
181-1557 181-0016 181-0045 181-1082 VACU20731102 181-0476 VACU20730002 VACU20743002 VACU20743001 VACU20741002 181-0477 181-0478 181-0437
Diaphragm pumps
Pumps Vacuum Diaphragm Pumps
These mechanical diaphragm pumps function without oil. They are easy to operate, use no water and generate neither wastewater nor used oil. These pumps are used for evacuating or pumping gases between containers in chemistry or physics laboratories and are extremely versatile.
  • Available in aluminium or chemical
  • Large selection of final pressure and pump capacities, high pump volume capacities even at low pressures
  • Maintenance-free drive unit, constructed for easy maintenance, diaphragm and valves are easy to replace
  • Very quiet, low-vibration
  • Optimised
More Product Information