Vacuum aspiration systems, VACUSAFE
Supplier: INTEGRA BIOSCIENCE
158320 158300 158310
391-2094EA 269500 INR
391-2094 181-0271 181-0270
Vacuum aspiration systems, VACUSAFE
Pumps Vacuum Aspiration Systems
These systems are used for aspirating, removing, or filtering biological or chemical liquids cleanly and efficiently. All functions necessary for this are built into the compact design. With single button operation, the device is ready for use at all times.
  • The vacuum system is always ready for use, fast, easy to operate
  • Helps to prevent contamination of the laboratory and the environment
  • Latest pump technology, ultra-quiet running
  • Pump switches off automatically when vacuum is reached
  • Easily transportable, independent of water supply and drainage system
More Product Information