VWR® VP 86, Mini Diaphragm Vacuum Pumps and Compressors
Supplier: VWR Collection
181-0067EA 42203.7 INR
181-0067 181-0065
VWR® VP 86, Mini Diaphragm Vacuum Pumps and Compressors
Pumps Vacuum Diaphragm Pumps
Single head, dry running diaphragm pump for a wide range of laboratory applications. It pumps, compresses and evacuates without affecting media, i.e., media remains uncontaminated. Includes diaphragm with patented tension optimisation, which allows the pump size to be smaller extending the service life of the diaphragm.
  • 100% oil-free pumping, so media is not contaminated when pumping, evacuating or compressing
  • For neutral to mildly aggressive gases and vapours
  • Maintenance-free, quiet running
* Flow rate at atmospheric pressure.
Delivery information:More Product Information