Isopropanol (Propan-2-ol), sterile spray
Supplier: ITW-CONTAMINATION TEXWIPE
TX3270
148-0025EA 0 INR
148-0025
Isopropanol (Propan-2-ol), sterile spray
Cleaning Agents Detergents and Surface Cleansers
Sterile 70% Propan-2-ol solution, pre-filtered.
  • Fully assembled trigger spray delivers fine stream or coarse spray
  • Gamma irradiated to a SAL (Sterility Assurance Level) of 10⁻⁶
  • With lot specific certificates of irradiation and analysis
Packaging: Double-bagged and packaged in sealed polybag case liner.