VWR®, Cable Ties, Polyamide 6.6
Supplier: VWR Collection
129-1607EA 0 INR
129-1607 129-1608 129-1609 129-1610 129-1611 129-1612 129-1613 129-1614 129-1615 129-1616 129-1617 129-1618 129-1619 129-1620 129-1621 129-1622
VWR®, Cable Ties, Polyamide 6.6
Ties
Pull-tight cable ties are available in different lengths ranging from 100 to 370 mm. All cable ties are made from polyamide 6.6 material.
  • High resistance to bases, oils, greases, and oil derivatives
  • Limited resistance to acids
  • Not resistant to phenols or chloride solvents
  • Operating temperature: −40 up to 85 °C
  • Installation temperature: −10 up to 60 °C
  • Melting point: 260 °C
  • Flammability class V2 acc- a UL 94
Easy to use in a wide set of applications, cable ties provide secure fastening solutions. A variety of colours allows users to identify the rightMore Product Information