Mops, PolyGen™
Supplier: MICRONOVA
PG9-12
129-0167EA 0 INR
129-0167
Mops, PolyGen™
Mops
The dual-action abrasive mesh is specially adapted to pharmaceutical and biotech projects. Heads used directly with the fibreglass/PP handle or attached to the universal handle via the quick disconnect adapter.
  • Mop can clean to a dry finish or apply disinfectants requiring extended drying time
  • Special herringbone weave ensures no raw edges to guarantee low particulation
  • Low-linting polyester looped strands
  • Reusable, launderable, and autoclavable
Ordering information: Mop heads and handle must be ordered separately.