VWR® Sharps Container Systems, Red
Supplier: VWR Collection
19001-010 19001-001 19001-013 19001-016 19001-020 19001-005 19001-006 19001-028 19001-030 19001-026 19001-023 19001-025 19001-008
119-0904EA 0 INR
119-0904 119-0900 119-0901 119-0905 119-0906 119-0903 119-0908 119-0910 119-0911 119-0909 119-0093 119-0094 119-0091
VWR® Sharps Container Systems, Red
Bins Sharps Containers
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
  • Puncture-resistant and autoclave- and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and autoclave- and incinerator-safe. They feature transparent, locking lids, and needle removal ports. The tortuous path systems restrict hand entry.
The extended neck sharps containersMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable