Swabs, Microdenier
Supplier: ITW-CONTAMINATION TEXWIPE
TX761MD
115-1899EA 1109.36 INR
115-1899
Swabs, Microdenier
Swabs Swabs for Cleaning
Swab series made from the cleanest microdenier material, 100% polyester, nylon-free. The complete thermal bond construction eliminates adhesive contamination.
  • Lot coded for traceability and quality control
  • 100% virgin polypropylene handle
Cleanroom laundered, providing ultra-low levels of particles, NVRs (non volatile residues) and ions, and engineered to provide enhanced absorbency and quick wicking performance. Streak-free cleaning with scratch-sensitive surfaces and compatible with QAC (quaternary ammonium chloride) cleaners and disinfectants.More Product Information