Safety winter boots, pull-on, uvex quatro pro, 8403
Supplier: UVEX
8403.2/40 8403.2/42 8403.2/43 8403.2/44 8403.2/45 8403.2/46 8403.2/41 8403.2/48 8403.2/38 8403.2/39 8403.2/47
UVEX8403.2/40EA 0 INR
UVEX8403.2/40 UVEX8403.2/42 114-7082 114-7083 114-7084 114-7085 UVEX8403.2/41 114-7087 UVEX8403.2/38 UVEX8403.2/39 114-7086
Safety winter boots, pull-on, uvex quatro pro, 8403
Shoes and Boots
Sturdy, comfortable anti-static safety boots with inside zip made from water resistant leather upper, soft padded collar and padded dust tongue and a dual-density, self-cleaning and slip-resistant PUR sole (4,5 mm, heat resistant to 120 °C, oil and petrol resistant, silicone-free).
  • Climazone technology for improved comfort
  • Warm fleece lining
  • Removable, anti-static insole with a moisture control system creates a superior foot climate and improved comfort
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the
More Product Information