VWR® Basic Mob Caps, Detectable
Supplier: VWR Collection
113-8895EA 0 INR
113-8895 113-8896 113-8897 113-8898
VWR® Basic Mob Caps, Detectable
Disposable Garments
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Double elastic
  • Metal detectable, therefore, ideal for use in the food processing industry
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.