VWR® Cleanroom Gloves, Nitrile, Sterile, ISO 5, 400 mm
Supplier: VWR Collection
112-5129EA 35409 INR
112-5129 112-5126 112-5125 112-5127 112-5128 112-5130
VWR® Cleanroom Gloves, Nitrile, Sterile, ISO 5, 400 mm
Gloves
These soft nitrile gloves are manufactured from carboxylated acrylonitrile butadiene that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Elbow length protection
  • Ambidextrous, micro textured fingers and palm, beaded cuff
  • Without natural rubber latex
  • Static-disspative
  • Single-use only
Recommended for ISO Class 5 (FED-STD-209E Class 100) or higher cleanrooms.
Certifications: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; AQL 1,5
Packaging: Gloves are packed 1 pair/pack, 10 packs/More Product Information