VWR®, Glove Liners
Supplier: VWR Collection
94000-608
VWRU94000-608EA 0 INR
VWRU94000-608 112-1898 112-1899 112-1900 112-1904 112-1905 112-1910 112-1915 112-1917 112-4326
VWR®, Glove Liners
Gloves
These durable and reusable glove liners are made from 100% nylon. They are compatible with stringent cleanroom requirements, and can be used effectively as a gowning glove. Low linting.
  • Reduces perspiration and wicks moisture during extended use
  • Prevents skin irritation without diminishing tactile sensitivity
  • Limits wear and tear to extend life of gloves, and conserve resources
  • Pre-laundered, will not shrink during washing
Basic weight: 21,5 g/pair ±10%
Thickness: 2 mm ±10%
Surface resistivity: 1×10⁹ ±10% ohms/m²
Used glove liners can beMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous / hand-specific Hand-specific
Disposable / reusable Reusable
Cuff style Knit wrist