Safety helmets, uvex pheos alpine
Supplier: UVEX
9773.350
111-2277EA 0 INR
111-2277
Safety helmets, uvex pheos alpine
Head Protection Safety Helmets
This light ABS helmet offers a high level of protection and durability. It is multifunctional and combines features from industrial protection helmets (EN 397) and requirements for climbing helmets (EN 12492).
  • Side Euroslot adapters (30 mm) for attaching earmuffs
  • Attachment for face protection available
  • Six-point textile suspension harness ensures optimum fit and comfort
  • Clips for attaching extras with elastic band, e.g. wide vision goggles and LED climbing helmet torch
  • Head torches (miner’s lamp) can be attached
  • Suitable for ambient temperatures down to −3
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable