Face shields, Clearways
Supplier: HONEYWELL SAFETY PRODUCTS
1002353 1002359 1002366
111-1254EA 0 INR
111-1254 111-1359 111-1360
Face shields, Clearways
Face Shields
This combination offers both strength and durability. The visors are suitable for different kinds of applications.
  • Lightweight - comfortable enough to wear all day
  • Integral visor fitting ensures extra protection against chemical seepage
  • Economical replaceable visor available in acetate or PC (PC: High impact; acetate: Chemical resistant)
Weight: 160 g
Visor: 200 mm
EN 166
Certifications: EN 166
Ordering information: Visor, brow guard, and chin guard must be ordered separately. See Accessories tab for full component listings. Holder mustMore Product Information