N,N-Dimethylacetamide 99,5, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

103646EEA 0 INR
103646E
N,N-Dimethylacetamide 99,5, AnalaR NORMAPUR® zA
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1 III
REACH: 01-2119459339-27
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Insolubility in water Passes test
IR Spectrum Passes test
Acidity Max. 0.02 meq/g
Colouration Max. 10 APHA
Non-volatile residue Max. 100 ppm
Water Max. 0.1 %
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 10 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR