Chloramphenicol 97,0-103,0, Ultra Pure Grade

Supplier: VWR Chemicals
Danger

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

0230-EU-100GEA 10805 INR
0230-EU-100G
Chloramphenicol 97,0-103,0, Ultra Pure Grade
Chloramphenicol

Inhibits protein synthesis at peptidyltransferase. Recommended working concentration: 20 μg/ml.

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Boiling Pt: 645
Melting Pt: 150
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

Specification Test Results

Chromatographic Purity PASS
Crystallinity PASS
Identification Pass
Melting Range 149 – 153 °C
pH (2.5%, Water) @ 25 °C 4.5 – 7.5
Purity 97.0 – 103.0 %
Specific Rotation +17.0 – +20.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR