Masterflex® Transfer Tubing, Convoluted FEP, Avantor®
Supplier: Avantor Fluid Handling
MFLX06486-04EA 23857.78 INR
MFLX06486-04 MFLX06486-08
Masterflex® Transfer Tubing, Convoluted FEP, Avantor®
Tubing Transfer Tubing
Promotes full drainage.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Convoluted spiral design promotes full drainage and increases tubing flexibility. This chemical-resistant fluoropolymer tubing is sold in 5-ft lengths with nonconvoluted 1"L sections at both ends for easy connections.